(1S,2S)-2-Fluorocyclopropanecarboxylic acid 95%
Supplier: Ambeed
|
Danger
|
A103810-100MG
A103810-1G
A103810-250MG
A103810-25G
A103810-5G
76982-332EA
18.04
USD
76982-332
76982-334
76982-336
77548-968
77565-800
(1S,2S)-2-Fluorocyclopropanecarboxylic acid 95%
(1S,2S)-2-Fluorocyclopropanecarboxylic acid
Precaución: For research use only.
Formula:
C₄H₅FO₂ MW: 104.08 g/mol |
MDL Number:
MFCD19237459 CAS Number: 127199-14-8 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...