Tyrphostin AG 1478 98%

Supplier: Ambeed
Warning

A103804-10MG A103804-1MG A103804-50MG A103804-5MG
77025-316EA 57.01 USD
77025-316 77025-318 77025-320 77025-322
Tyrphostin AG 1478 98%
Tyrphostin AG 1478

Precaución: for Research Use Only

Formula: C₁₆H₁₄ClN₃O₂
MDL Number: MFCD00270914
CAS Number: 153436-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR