2-Methyl-2H-indazole-4-boronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

A103767-100MG A103767-1G A103767-250MG A103767-5G
77025-306EA 9 USD
77025-306 77025-308 77025-310 77025-312
2-Methyl-2H-indazole-4-boronic acid pinacol ester 96%
2-Methyl-2H-indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD11109437
CAS Number: 885698-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR