2'-O-Methylcytidine 97%

Supplier: Ambeed
Warning

Synonyms: 2′-O-methylcytidine

A103746-5G A103746-1G A103746-25G A103746-10G A103746-250MG A103746-100G
76994-662EA 82.76 USD
76994-662 77565-796 77570-214 77599-508 77599-510 77670-504
2'-O-Methylcytidine 97%
2'-O-Methylcytidine

Precaución: For research use only.

Formula: C₁₀H₁₅N₃O₅
MW: 257.25 g/mol
MDL Number: MFCD00056067
CAS Number: 2140-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR