4,7,10,13-Tetraoxahexadecane-1,16-dinitrile 98%

Supplier: Ambeed
Warning

A103671-100MG A103671-1G A103671-250MG
77008-502EA 39 USD
77008-502 77008-504 77008-506
4,7,10,13-Tetraoxahexadecane-1,16-dinitrile 98%
4,7,10,13-Tetraoxahexadecane-1,16-dinitrile

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₄
MDL Number: MFCD00483880
CAS Number: 57741-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR