3-Bromo-4-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A103591-100G A103591-10G A103591-1G A103591-250MG A103591-25G A103591-5G
77008-490EA 1330.37 USD
77008-490 77008-492 77008-494 77008-496 77008-498 77008-500
3-Bromo-4-nitrobenzoic acid 97%
3-Bromo-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
MDL Number: MFCD04117949
CAS Number: 101420-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR