4-Iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A103500-1G
A103500-25G
A103500-5G
A103500-100G
77008-484EA
6.37
USD
77008-484
77008-486
77008-488
77682-948
4-Iodopyridine 98%
4-Iodopyridine
Precaución: For research use only.
Formula:
C₅H₄IN MW: 205 g/mol |
MDL Number:
MFCD02093937 CAS Number: 15854-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...