1-Methyl-1H-indole-6-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-Cyano-1-methylindole

A103498-10G A103498-1G A103498-250MG A103498-5G
77025-294EA 249.32 USD
77025-294 77025-296 77025-298 77025-300
1-Methyl-1H-indole-6-carbonitrile 97%
1-Methyl-1H-indole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈N₂
MDL Number: MFCD08690254
CAS Number: 20996-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR