β-(N-Morpholino)ethanesulfonic acid (MES) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
A103489-1000G
A103489-100G
A103489-10G
A103489-25G
A103489-500G
77025-284EA
158.08
USD
77025-284
77025-286
77025-288
77025-290
77025-292
β-(N-Morpholino)ethanesulfonic acid (MES) 98%
β-(N-Morpholino)ethanesulphonic acid (MES)
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₄S MW: 195.24 g/mol |
MDL Number:
MFCD00006181 CAS Number: 4432-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...