PJ-34 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride, PJ34 HCl

A103485-100MG A103485-10MG A103485-1MG A103485-250MG A103485-50MG
76982-294EA 121 USD
76982-294 76982-296 76982-298 76982-300 76982-302
PJ-34 hydrochloride 98%
PJ-34 hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₇N₃O₂·HCl
MDL Number: MFCD22581510
CAS Number: 344458-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR