1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1,4,5,6-Tetrahydro-pyrrolo[3,4-c]pyrazole,hydrochloride

A103473-1G A103473-250MG A103473-5G
77025-278EA 32 USD
77025-278 77025-280 77025-282
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₉Cl₂N₃
MDL Number: MFCD08457551
CAS Number: 157327-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR