(R)-1-(p-Tolyl)butan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A103467-100MG A103467-1G A103467-250MG
76982-288EA 15.92 USD
76982-288 76982-290 76982-292
(R)-1-(p-Tolyl)butan-1-amine hydrochloride 95%
(R)-1-(p-Tolyl)butan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈ClN
MW: 199.72 g/mol
MDL Number: MFCD12757109
CAS Number: 851597-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR