(2-Bromo-5-chloropyridin-3-yl)methanol 98%

Supplier: Ambeed
Warning

A103428-1G A103428-250MG
77008-480EA 333.13 USD
77008-480 77008-482
(2-Bromo-5-chloropyridin-3-yl)methanol 98%
(2-Bromo-5-chloropyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.47 g/mol
MDL Number: MFCD16606984
CAS Number: 1227585-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR