3-(2-Chlorobenzyloxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-((2-Chlorobenzyl)oxy)phenyl)boronic acid

A103410-1G A103410-250MG A103410-5G
77008-474EA 64.72 USD
77008-474 77008-476 77008-478
3-(2-Chlorobenzyloxy)phenylboronic acid 98%
3-(2-Chlorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MW: 262.5000 g/mol
MDL Number: MFCD05865198
CAS Number: 845551-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR