[6]-Gingerol 98%

Supplier: Ambeed
Danger

Synonyms: (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)decan-3-one, 1-(4ʹ-Hydroxy-3ʹ-methoxyphenyl)-5-hydroxy-3-decanone, 6-Gingerol, (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, Gingerol

A103354-10MG A103354-1MG A103354-5MG A103354-250MG A103354-100MG
77025-266EA 47.75 USD
77025-266 77025-268 77025-270 77599-502 77666-494
[6]-Gingerol 98%
[6]-Gingerol

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₄
MW: 294.4 g/mol
MDL Number: MFCD00210507
CAS Number: 23513-14-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR