4,6-Dichloro-2-fluoropyrimidine 95%

Supplier: Ambeed
Warning

A103341-100MG A103341-1G
77008-472EA 50 USD
77008-472 77889-722
4,6-Dichloro-2-fluoropyrimidine 95%
4,6-Dichloro-2-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₂FN₂
MDL Number: MFCD27939250
CAS Number: 3824-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR