2-Amino-4,6-dichloro-5-methylpyrimidine 95%

Supplier: Ambeed
Warning

A103329-100MG A103329-10G A103329-1G A103329-250MG A103329-25G A103329-5G
77008-460EA 7 USD
77008-460 77008-462 77008-464 77008-466 77008-468 77008-470
2-Amino-4,6-dichloro-5-methylpyrimidine 95%
2-Amino-4,6-dichloro-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N₃
MDL Number: MFCD00023253
CAS Number: 7153-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR