7-Chlorooxindole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Chloroindolin-2-one
A103224-1G
A103224-250MG
A103224-5G
76994-638EA
29.71
USD
76994-638
76994-640
76994-642
7-Chlorooxindole 97%
7-Chlorooxindole
Precaución: for Research Use Only
Formula:
C₈H₆ClNO MW: 167.5923 g/mol |
MDL Number:
MFCD06656001 CAS Number: 25369-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...