3-Benzyloxy-5-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyloxy-5-fluorobenzeneboronic acid

A103165-1G A103165-250MG
77025-260EA 22 USD
77025-260 77025-262
3-Benzyloxy-5-fluorophenylboronic acid 97%
3-Benzyloxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD07363782
CAS Number: 850589-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR