Ethyl 5-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

A103162-100MG A103162-250MG
77008-446EA 15 USD
77008-446 77008-448
Ethyl 5-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate 95%
Ethyl 5-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
CAS Number: 85614-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR