Methyl 2,6-dichloro-4-methylnicotinate 98%

Supplier: Ambeed
Warning

A103107-10G A103107-1G A103107-250MG A103107-5G
77025-242EA 847.66 USD
77025-242 77025-244 77025-246 77025-248
Methyl 2,6-dichloro-4-methylnicotinate 98%
Methyl 2,6-dichloro-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
MDL Number: MFCD15144593
CAS Number: 1013648-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR