Econazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-((4-Chlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole, 1-[2,4-Dichloro-beta-[(p-chlorobenzyl)oxy]phenethyl]imidazole

A103065-10G A103065-25G
77025-232EA 18.04 USD
77025-232 77025-234
Econazole 97%
Econazole

Precaución: for Research Use Only

Formula: C₁₈H₁₅Cl₃N₂O
MW: 381.69 g/mol
MDL Number: MFCD00800993
CAS Number: 27220-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR