3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-hydroxybenzamide 98%

Supplier: Ambeed
Warning

A103031-1G A103031-250MG
76982-272EA 1687 USD
76982-272 76982-274
3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-hydroxybenzamide 98%
3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-hydroxybenzamide

Precaución: for Research Use Only

Formula: C₁₆H₁₄Cl₂N₂O₃
MDL Number: MFCD23703091
CAS Number: 475271-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR