6-Bromo-1-methyl-1H-benzo[d]imidazol-2(3H)-one 95%

Supplier: Ambeed
Warning

A103029-1G A103029-25G A103029-5G A103029-100MG A103029-250MG
77008-432EA 16 USD
77008-432 77008-434 77008-436 77889-719 77889-720
6-Bromo-1-methyl-1H-benzo[d]imidazol-2(3H)-one 95%
6-Bromo-1-methyl-1H-benzo[d]imidazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MDL Number: MFCD09759338
CAS Number: 305790-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR