(S)-tert-Butyl (2-aminopropyl)carbamate 95%

Supplier: Ambeed
Warning

A103000-100MG A103000-10G A103000-250MG A103000-1G A103000-5G
76994-616EA 27 USD
76994-616 76994-618 76994-620 77670-482 77670-484
(S)-tert-Butyl (2-aminopropyl)carbamate 95%
(S)-tert-Butyl (2-aminopropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD11112235
CAS Number: 121103-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR