6,6'-(9H-Fluorene-9,9-diyl)bis(2-naphthol) 95%

Supplier: Ambeed
Warning

Synonyms: 9-Bis(6-hydroxy-2-naphthyl)fluorene, 6,6'-(9H-Fluorene-9,9-diyl)bis(naphthalen-2-ol)

A102980-100G A102980-10G A102980-1G A102980-250MG A102980-25G A102980-5G
76982-260EA 140 USD
76982-260 76982-262 76982-264 76982-266 76982-268 76982-270
6,6'-(9H-Fluorene-9,9-diyl)bis(2-naphthol) 95%
6,6'-(9H-Fluorene-9,9-diyl)bis(2-naphthol)

Precaución: for Research Use Only

Formula: C₃₃H₂₂O₂
MDL Number: MFCD24038938
CAS Number: 934557-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR