3,6-Dihydrothiopyran-4-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran, 2-(3,6-Dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A102956-1G A102956-250MG A102956-5G
76982-254EA 45.62 USD
76982-254 76982-256 76982-258
3,6-Dihydrothiopyran-4-boronic acid pinacol ester 97%
3,6-Dihydrothiopyran-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BO₂S
MW: 226.141 g/mol
MDL Number: MFCD10700154
CAS Number: 862129-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR