3-Chloro-2-nitrobenzonitrile 96%

Supplier: Ambeed
Warning

A102919-100MG A102919-1G A102919-250MG A102919-25G A102919-5G
77008-418EA 10.61 USD
77008-418 77008-420 77008-422 77008-424 77008-426
3-Chloro-2-nitrobenzonitrile 96%
3-Chloro-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MW: 182.564 g/mol
MDL Number: MFCD15529744
CAS Number: 34662-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR