3-Amino-6-bromo-2-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine

A102903-10G A102903-1G A102903-250MG A102903-25G A102903-5G
76994-602EA 33.95 USD
76994-602 76994-604 76994-606 76994-608 76994-610
3-Amino-6-bromo-2-methylpyridine 98%
3-Amino-6-bromo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD03095086
CAS Number: 126325-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR