4-Methyl-4-(methylthio)-2-pentanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-4-methylthio-2-pentanone, 4-Methyl-4-(methylthio)pentan-2-one

A102876-100G A102876-25G A102876-5G
76994-596EA 78.51 USD
76994-596 76994-598 76994-600
4-Methyl-4-(methylthio)-2-pentanone 98%
4-Methyl-4-(methylthio)-2-pentanone

Precaución: for Research Use Only

Formula: C₇H₁₄OS
MW: 146.25 g/mol
MDL Number: MFCD00049253
CAS Number: 23550-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR