2,2,2-Trifluoro-1-(p-tolyl)ethanol 98%

Supplier: Ambeed
Warning

A102869-100MG A102869-1G A102869-250MG A102869-5G
76982-240EA 17 USD
76982-240 76982-242 76982-244 76982-246
2,2,2-Trifluoro-1-(p-tolyl)ethanol 98%
2,2,2-Trifluoro-1-(p-tolyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MDL Number: MFCD00233111
CAS Number: 446-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR