2,2-Bis[4-(4-aminophenoxy)phenyl]propane 97%

Supplier: Ambeed
Warning

A102792-100G A102792-1000G A102792-500G
76994-590EA 36 USD
76994-590 77548-940 77880-834
2,2-Bis[4-(4-aminophenoxy)phenyl]propane 97%
2,2-Bis[4-(4-aminophenoxy)phenyl]propane

Precaución: for Research Use Only

Formula: C₂₇H₂₆N₂O₂
MDL Number: MFCD00039152
CAS Number: 13080-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR