Baricitinib 99%

Supplier: Ambeed
Warning

Synonyms: LY-3009104, INCB028050, LY3009104, 2-[1-Ethylsulphonyl-3-[4-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)pyrazol-1-yl]azetidin-3-yl]acetonitrile, 2-(3-(4-(3H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-1-(ethylsulphonyl)azetidin-3-yl)acetonitrile, LY 3009104, INCB-028050, INCB 028050

A102707-10MG A102707-1G A102707-1MG A102707-250MG A102707-2MG A102707-5G A102707-5MG A102707-100MG
77008-392EA 35.01 USD
77008-392 77008-394 77008-396 77008-398 77008-400 77008-402 77008-404 77682-945
Baricitinib 99%
Baricitinib

Precaución: For research use only.

Formula: C₁₆H₁₇N₇O₂S
MW: 371.42 g/mol
MDL Number: MFCD27920779
CAS Number: 1187594-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR