2,6-Dichloro-N,N-dimethylpyridin-4-amine 98%

Supplier: Ambeed
Warning

A102683-100MG A102683-250MG A102683-1G
77008-388EA 27.59 USD
77008-388 77008-390 77610-774
2,6-Dichloro-N,N-dimethylpyridin-4-amine 98%
2,6-Dichloro-N,N-dimethylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂
MW: 191.06 g/mol
MDL Number: MFCD09031039
CAS Number: 175461-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR