4-(Hydroxymethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: (1H-Indazol-4-yl)methanol

A102644-100MG A102644-1G A102644-250MG A102644-25G A102644-5G
77025-216EA 7.43 USD
77025-216 77025-218 77025-220 77025-222 77025-224
4-(Hydroxymethyl)-1H-indazole 95%
4-(Hydroxymethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD11052642
CAS Number: 709608-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR