5-Methoxy-7-azaindole 98%

Supplier: Ambeed
Warning

A102598-100MG A102598-1G A102598-250MG A102598-25G A102598-5G A102598-100G
77008-380EA 7 USD
77008-380 77008-382 77008-384 77008-386 77570-148 77889-717
5-Methoxy-7-azaindole 98%
5-Methoxy-7-azaindole

Precaución: for Research Use Only

Formula: C8H8N2O
MDL Number: MFCD06659675
CAS Number: 183208-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR