7-Bromo-5-(trifluoromethyl)-1H-indazole 98%

Supplier: Ambeed
Warning

A102573-100MG A102573-250MG
77008-376EA 38.2 USD
77008-376 77008-378
7-Bromo-5-(trifluoromethyl)-1H-indazole 98%
7-Bromo-5-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD27939136
CAS Number: 1100212-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR