(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride 95%

Supplier: Ambeed
Warning

A102342-100MG A102342-1G A102342-250MG
76982-224EA 45 USD
76982-224 76982-226 76982-228
(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride 95%
(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂FNO·ClH
MDL Number: MFCD16294167
CAS Number: 1149383-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR