2-(Methylthio)pyrimidine-4,6-diol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylsulphanyl-pyrimidine-4,6-diol, 4,6-Dihydroxy-2-methylsulphanylpyrimidine

A102341-1000G A102341-100G A102341-10G A102341-25G A102341-500G A102341-5G
77008-364EA 183.54 USD
77008-364 77008-366 77008-368 77008-370 77008-372 77008-374
2-(Methylthio)pyrimidine-4,6-diol 97%
2-(Methylthio)pyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.18 g/mol
MDL Number: MFCD00006087
CAS Number: 1979-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR