Benzyl 3-methyl-4-oxopiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A102285-1G A102285-5G
76982-222EA 25.47 USD
76982-222 77570-124
Benzyl 3-methyl-4-oxopiperidine-1-carboxylate 98%
Benzyl 3-methyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD11848402
CAS Number: 1010115-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR