Adipohydrazide 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Adipyl hydrazide
A102141-100G
A102141-500G
76994-570EA
17
USD
76994-570
76994-572
Adipohydrazide 95%
Adipohydrazide
Precaución: for Research Use Only
Formula:
C₆H₁₄N₄O₂ MW: 174.2 g/mol Boiling Pt: 446 |
MDL Number:
MFCD00007614 CAS Number: 1071-93-8 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...