4-(Hydroxymethyl)-2,6-dimethylphenol 98%

Supplier: Ambeed
Warning

A101979-100MG A101979-1G A101979-250MG
76994-554EA 27.59 USD
76994-554 76994-556 76994-558
4-(Hydroxymethyl)-2,6-dimethylphenol 98%
4-(Hydroxymethyl)-2,6-dimethylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD02912490
CAS Number: 4397-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR