a-Tosyl(2-bromobenzyl)isocyanide 95%

Supplier: Ambeed
Warning

A101945-1G A101945-250MG
77008-350EA 139 USD
77008-350 77008-352
a-Tosyl(2-bromobenzyl)isocyanide 95%
a-Tosyl(2-bromobenzyl)isocyanide

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrNO₂S
MDL Number: MFCD04114784
CAS Number: 936548-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR