Ethyl 7-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

A101932-100MG A101932-250MG A101932-50MG
77025-180EA 112 USD
77025-180 77025-182 77025-184
Ethyl 7-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylate 95%
Ethyl 7-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₃
MW: 296.11 g/mol
MDL Number: MFCD09746289
CAS Number: 208580-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR