Benzyl (4-(2-bromoacetyl)phenyl)carbamate 95%

Supplier: Ambeed
Danger

A101392-10G A101392-1G A101392-250MG A101392-5G
77008-324EA 487 USD
77008-324 77008-326 77008-328 77008-330
Benzyl (4-(2-bromoacetyl)phenyl)carbamate 95%
Benzyl (4-(2-bromoacetyl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₆H₁₄BrNO₃
MW: 348.19 g/mol
MDL Number: MFCD05663954
CAS Number: 157014-41-0
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR