2-(Methylsulfonyl)-4-(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A100777-100G A100777-10G A100777-1G A100777-250MG A100777-25G A100777-5G
77025-144EA 867 USD
77025-144 77025-146 77025-148 77025-150 77025-152 77025-154
2-(Methylsulfonyl)-4-(trifluoromethyl)benzoic acid 95%
2-(Methylsulphonyl)-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₄S
MDL Number: MFCD09037774
CAS Number: 142994-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR