2-(4-Amino-3-methylphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A100569-1G A100569-250MG A100569-100MG
77008-280EA 290.01 USD
77008-280 77008-282 77891-746
2-(4-Amino-3-methylphenyl)acetic acid 98%
2-(4-Amino-3-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD11036405
CAS Number: 705240-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR