Noopept 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Ethyl 2-(1-(2-phenylacetyl)pyrrolidine-2-carboxamido)acetate, N-[1-(Phenylacetyl)-L-prolyl]glycine Ethyl ester, Omberacetam, Ethyl N-[1-(Phenylacetyl)-L-prolyl]glycinate

A100439-100MG A100439-1G A100439-25G A100439-5G A100439-100G
76982-130EA 5.31 USD
76982-130 76982-132 76982-134 76982-136 77682-931
Noopept 98%
Noopept

Precaución: For research use only.

Formula: C₁₇H₂₂N₂O₄
MW: 318.37 g/mol
MDL Number: MFCD00941121
CAS Number: 157115-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR