Ethyl 5-bromopyrimidine-4-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A100428-100MG
A100428-250MG
A100428-5G
A100428-1G
76994-444EA
21.22
USD
76994-444
76994-446
76994-448
77670-448
Ethyl 5-bromopyrimidine-4-carboxylate 98%
Ethyl 5-bromopyrimidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₇BrN₂O₂ MW: 231.04 g/mol |
MDL Number:
MFCD16660482 CAS Number: 64224-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...