3-Amino-5-methylpyrazine-2-carbonitrile 95%

Supplier: Ambeed
Danger

A100331-100MG A100331-1G
77008-262EA 140.04 USD
77008-262 77008-264
3-Amino-5-methylpyrazine-2-carbonitrile 95%
3-Amino-5-methylpyrazine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₆N₄
MW: 134.13 g/mol
CAS Number: 58091-66-0
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR